Learn More
Ace Glass, Inc. Lab bottle, Duran, 15L, plain, pouring ring & GL 45 PP bluecap, cs/1
SDP
Catalog No. 50978382
Encompass
Chemical-resistant and stable 15mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1 aEc15000mL (507oz) CapacityaEc277mm DiameteraEc450mm Height w/Cap
Catalog No. 50-978-382 Supplier Ace Glass, Inc. Supplier No. 553990
Chemical-resistant and stable 15mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1 aEc15000mL (507oz) CapacityaEc277mm DiameteraEc450mm Height w/Cap