Learn More
Ace Glass, Inc. Lab bottle, Duran, 15000mL, plain GL 45, no cap or pour ring, cs/1
SDP
Catalog No. 50978379
Encompass
Chemical-resistant and stable 15L Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with GL45 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/1.
Catalog No. 50-978-379 Supplier Ace Glass, Inc. Supplier No. 553968
Chemical-resistant and stable 15L Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with GL45 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/1.