Learn More
Ace Glass, Inc. Lab bottle, Duran, 15L, plain, pouring ring & PTFE lined GL 45PBT cap, cs/1
SDP
Catalog No. 50971910
Encompass
Chemical-resistant and stable 15L Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1.
Catalog No. 50-971-910 Supplier Ace Glass, Inc. Supplier No. 553941
Chemical-resistant and stable 15L Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1.