Learn More
Ace Glass, Inc. Lab bottle, Duran, 1L, GL 45, amber, pressure plus, cs/10
SDP
Catalog No. 50971963
Encompass
Chemical-resistant and stable, 1L amber laboratory bottle, approved for vacuum or pressure use. Rated at -1 to +1.5bar (-14.5 to +21.7PSI). Supplied with blue graduations. Does not include cap or pouring ring. Temperature range up to 140 deg.C.
Catalog No. 50-971-963 Supplier Ace Glass, Inc. Supplier No. 5557121
Chemical-resistant and stable, 1L amber laboratory bottle, approved for vacuum or pressure use. Rated at -1 to +1.5bar (-14.5 to +21.7PSI). Supplied with blue graduations. Does not include cap or pouring ring. Temperature range up to 140 deg.C.