Learn More
Ace Glass, Inc. Lab bottle, Duran, 1000mL, absolute vacuum to 1.5 bar gauge, netted, pouring ring & blue GL 45 polypropylene cap, cs/1
SDP
Catalog No. 50971943
Encompass
Chemical-resistant and stable 1L Duran laboratory bottles, fabricated from borosilicate glass and netted. Supplied graduated with PTFE-lined GL45 PP cap and pouring ring. Suitable for vacuum or pressure use up to 14psi. Case/1.
Catalog No. 50-971-943 Supplier Ace Glass, Inc. Supplier No. 5539224
Chemical-resistant and stable 1L Duran laboratory bottles, fabricated from borosilicate glass and netted. Supplied graduated with PTFE-lined GL45 PP cap and pouring ring. Suitable for vacuum or pressure use up to 14psi. Case/1.