Learn More
Ace Glass, Inc. Lab bottle, Duran, 20L, amber, GL 45, no ring or cap, cs/1
SDP
Catalog No. 50971955
Encompass
Chemical-resistant and stable 20L Duran laboratory bottles, fabricated from borosilicate glass and ambered. Supplied graduated with GL45 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/1.
Catalog No. 50-971-955 Supplier Ace Glass, Inc. Supplier No. 5539268
Chemical-resistant and stable 20L Duran laboratory bottles, fabricated from borosilicate glass and ambered. Supplied graduated with GL45 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/1.