Learn More
Ace Glass, Inc. Lab bottle, Duran, 250mL, absolute vacuum to 1.5 bar gauge, netted, pouring ring & blue GL 45 polypropylene cap, cs/1
SDP
Catalog No. 50971940
Encompass
Chemical-resistant and stable 250mL Duran laboratory bottles, fabricated from borosilicate glass and netted. Supplied graduated with PTFE-lined GL45 PP cap and pouring ring. Suitable for vacuum or pressure use up to 14psi. Case/1.
Catalog No. 50-971-940 Supplier Ace Glass, Inc. Supplier No. 5539216
Chemical-resistant and stable 250mL Duran laboratory bottles, fabricated from borosilicate glass and netted. Supplied graduated with PTFE-lined GL45 PP cap and pouring ring. Suitable for vacuum or pressure use up to 14psi. Case/1.