Learn More
Ace Glass, Inc. Lab bottle, Duran, 25mL, plain, GL 25 PP blue cap, no pouring ring, cs/10
SDP
Catalog No. 50978380
Encompass
Chemical-resistant and stable 25mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL25 PP cap. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-978-380 Supplier Ace Glass, Inc. Supplier No. 553981
Chemical-resistant and stable 25mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL25 PP cap. Can be hot-air sterilized or autoclaved. Case/10.