Learn More
Ace Glass, Inc. Lab bottle, Duran, 25mL, plain, PTFE lined GL 25 PBT cap, cs/10
SDP
Catalog No. 50971904
Encompass
Chemical-resistant and stable 25mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL25 PBT cap. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-971-904 Supplier Ace Glass, Inc. Supplier No. 553905
Chemical-resistant and stable 25mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL25 PBT cap. Can be hot-air sterilized or autoclaved. Case/10.