Learn More
Ace Glass, Inc. Lab bottle, Duran, 2L, plain, pouring ring & PTFE lined GL 45PBT cap, cs/10
SDP
Catalog No. 50971908
Encompass
Chemical-resistant and stable 2L Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-971-908 Supplier Ace Glass, Inc. Supplier No. 553925
Chemical-resistant and stable 2L Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.