Learn More
Ace Glass, Inc. Lab bottle, Duran, 2000mL, safety coated, GL 45, no pouring ring or cap, cs/10
SDP
Catalog No. 50978385
Encompass
Chemical-resistant and stable 2L Duran laboratory bottles, fabricated from borosilicate glass and safety coated. Supplied graduated with GL45 threads. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-978-385 Supplier Ace Glass, Inc. Supplier No. 5539158
Chemical-resistant and stable 2L Duran laboratory bottles, fabricated from borosilicate glass and safety coated. Supplied graduated with GL45 threads. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/10.