Learn More
Ace Glass, Inc. Lab bottle, Duran, 5000mL, plain, pouring ring & GL 45 PP bluecap, cs/1
SDP
Catalog No. 50971926
Encompass
Chemical-resistant and stable 5,000mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1.
Catalog No. 50-971-926 Supplier Ace Glass, Inc. Supplier No. 553988
Chemical-resistant and stable 5,000mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1.