Learn More
Ace Glass, Inc. Lab bottle, Duran, 500mL, plain, pouring ring & GL 45 PP bluecap, cs/10
SDP
Catalog No. 50971922
Encompass
Chemical-resistant and stable 500mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable GL45 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-971-922 Supplier Ace Glass, Inc. Supplier No. 553982
Chemical-resistant and stable 500mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable GL45 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.