Learn More
Ace Glass, Inc. Lab bottle, Duran, 50mL, plain GL 32, no cap or pour ring, cs/10
SDP
Catalog No. 50978377
Encompass
Chemical-resistant and stable 50mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with GL32 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-978-377 Supplier Ace Glass, Inc. Supplier No. 553952
Chemical-resistant and stable 50mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with GL32 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/10.