Learn More
Ace Glass, Inc. Lab bottle, Duran, 50mL, plain, pouring ring & GL 32 PP bluecap, cs/10
SDP
Catalog No. 50971923
Encompass
Chemical-resistant and stable 50mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL32 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-971-923 Supplier Ace Glass, Inc. Supplier No. 553983
Chemical-resistant and stable 50mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL32 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.