Learn More
Ace Glass, Inc. Lab bottle, Duran, 1L, GL 45, safety coated, pressure plus, cs/10
SDP
Catalog No. NC0493550
Encompass
Chemical-resistant and stable, 1L safety coated lab bottle, approved for vacuum or pressure use. Rated at -1 to +1.5bar (-14.5 to +21.7PSI). Supplied with blue graduations. Does not include cap or pour ring. Temperature range up to 140 deg.C.
Catalog No. NC0493550 Supplier Ace Glass, Inc. Supplier No. 5557214
Chemical-resistant and stable, 1L safety coated lab bottle, approved for vacuum or pressure use. Rated at -1 to +1.5bar (-14.5 to +21.7PSI). Supplied with blue graduations. Does not include cap or pour ring. Temperature range up to 140 deg.C.