Learn More
Chemglass Life Sciences Lab Clamp, Dual Adjust, Medium, Maximum Grip 60mm, Rod OD x Length (mm): 11 x 150
SDP
Catalog No. 501311235
Encompass_Preferred
Lab Clamp, Dual Adjust, Medium, Maximum Grip 60mm, Rod OD x Length (mm): 11 x 150• Dual adjusting knobs• High-strength, nickel-plated, zinc alloy• Vinyl-coated jawsClamps are constructed of high tensile-strength, nickel-plated, zinc alloy which will not rust. Two separate adjusting knobs ensure even pressure on glass to minimize breakage. Jaws are coated with blue vinyl for excellent grip.
Catalog No. 50-131-1235 Supplier Chemglass Life Sciences Supplier No. CG9004202
Lab Clamp, Dual Adjust, Medium, Maximum Grip 60mm, Rod OD x Length (mm): 11 x 150• Dual adjusting knobs• High-strength, nickel-plated, zinc alloy• Vinyl-coated jawsClamps are constructed of high tensile-strength, nickel-plated, zinc alloy which will not rust. Two separate adjusting knobs ensure even pressure on glass to minimize breakage. Jaws are coated with blue vinyl for excellent grip.