Learn More
Cole-Parmer Lab Companion AAHK34013U 9 Position Stirring Hot Plate, 120 V.
Catalog No. NC0870402 Shop All Cole-Parmer Products
Encompass_Preferred
These multiposition stirring hot plates have a smooth-start mechanism that prevents decoupling of stir bars even with viscous media. Featuring an easy-to-read and intuitive control panel with bright LED lights indicating stirring position and a heating operation light. Select from four operating modes: all, row, column, or individual. Control each mode with the built in timer for unattended operation. Over-temperature and current limit protection provide added user safety.
Integrated heater and aluminum top plate design provide exceptional heat up and transfer to samples. Powder-coated steel body design protects internal components from splashes and spills and attached non-slip silicone top plate mat prevents unintended slips.
Catalog No. NC0870402 Supplier Cole-Parmer Supplier No. 5146006
Add to Cart
Add to Cart
These multiposition stirring hot plates have a smooth-start mechanism that prevents decoupling of stir bars even with viscous media. Featuring an easy-to-read and intuitive control panel with bright LED lights indicating stirring position and a heating operation light. Select from four operating modes: all, row, column, or individual. Control each mode with the built in timer for unattended operation. Over-temperature and current limit protection provide added user safety.
Integrated heater and aluminum top plate design provide exceptional heat up and transfer to samples. Powder-coated steel body design protects internal components from splashes and spills and attached non-slip silicone top plate mat prevents unintended slips.