Learn More
Braintree Scientific Lab Control Unit - Single Gas Output, (2) presets
SDP
Catalog No. 501954732
Encompass
SMARTBOX™ Lab Control Unit Compliant with AVMA Guidelines Operates at a fixed flow rate based on the external CO2 source. The Controller automates on/off gas flow at the push of a button, delivering CO2 to individual cage or Euthanex lid. The EA-31000/32000 have a single gas output. The EA-32000 offers two preset times that are programmable. This economical unit is designed for small scale use by individual investigators in their laboratories. Lids and chambers not included! Lab Control Unit Features: Treats animals at individual cage level Designed for investigators that need to retrieve animals quickly Utilizing the full range of Euthanex Cage Lids or existing induction chamber Minimizes operator error and excessive gas use Automatic shut-off saves gas and protects personnel EA-32000 Double throw switches allow for two different time settings for the single output Easily attaches to wall **Sold for research purposes only and not for human use**
Catalog No. 50-195-4732 Supplier Braintree Scientific Supplier No. EA32000
SMARTBOX™ Lab Control Unit Compliant with AVMA Guidelines Operates at a fixed flow rate based on the external CO2 source. The Controller automates on/off gas flow at the push of a button, delivering CO2 to individual cage or Euthanex lid. The EA-31000/32000 have a single gas output. The EA-32000 offers two preset times that are programmable. This economical unit is designed for small scale use by individual investigators in their laboratories. Lids and chambers not included! Lab Control Unit Features: Treats animals at individual cage level Designed for investigators that need to retrieve animals quickly Utilizing the full range of Euthanex Cage Lids or existing induction chamber Minimizes operator error and excessive gas use Automatic shut-off saves gas and protects personnel EA-32000 Double throw switches allow for two different time settings for the single output Easily attaches to wall **Sold for research purposes only and not for human use**