Learn More
StatLab Lab Countertop Absorbant Sheets
Description
- Each sheet consists of four layers of wet-strength tissue, embossed together and adhesive laminated to a polypropylene moisture barrier
- Sheets are excellent for use in areas of routine and special staining and coverslipping, placing under a microtome, or in any other general lab application
Specifications
Specifications
| Material | Polypropylene |
| Type | Absorbent Pad |
| Disposable | Limited-use |
| Size | Large |
| Weight Capacity | Heavyweight |