Learn More
Alkali Scientific Lab Cryo Rectangle 1/4 x 3/4 White
SDP
Catalog No. 5000662083
Encompass_Preferred
The Lab Cryo Rectangle 1/4" x 3/4" White Labels are specialized cryogenic labels designed for precise identification and long term storage of samples in ultra low temperature environments. These labels are ideal for use with microtubes, cryovials, and other laboratory containers subjected to conditions such as liquid nitrogen, 80°C freezers, and dry ice.
Catalog No. 50-006-62083 Supplier Alkali Scientific Supplier No. LCR1434W
The Lab Cryo Rectangle 1/4" x 3/4" White Labels are specialized cryogenic labels designed for precise identification and long term storage of samples in ultra low temperature environments. These labels are ideal for use with microtubes, cryovials, and other laboratory containers subjected to conditions such as liquid nitrogen, 80°C freezers, and dry ice.