Learn More
Cole-Parmer IKA 3365000 Lab Dancer Compact Shaker, 2800 rpm, 100 to 240 VAC
Catalog No. NC0525419 Shop All Cole-Parmer Products
Encompass
Ideal for small vessels up to 30-mm diameter and 50-mL volume. Lightly press test tubes, centrifuge tubes, or Eppendorf vessels against the inert plastic test tube surface to activate shaker. Its economical, compact design makes it an indispensable tool for your laboratory.
Catalog No. NC0525419 Supplier Cole-Parmer Supplier No. 8400181
Ideal for small vessels up to 30-mm diameter and 50-mL volume. Lightly press test tubes, centrifuge tubes, or Eppendorf vessels against the inert plastic test tube surface to activate shaker. Its economical, compact design makes it an indispensable tool for your laboratory.