Learn More
Kebby Industries LAB DECRIMPER, 28MM W/ GRIPS
Catalog No. NC1481695
Encompass_Preferred
Laboratory Grade Decrimper tool for manual decrimping (decapping) of 28mm aluminum seals from vials. Supplied with cushioned grips.
Catalog No. NC1481695 Supplier Kebby Industries Supplier No. HCA2800100D05A
Laboratory Grade Decrimper tool for manual decrimping (decapping) of 28mm aluminum seals from vials. Supplied with cushioned grips.