Learn More
Research Products International Corp Laboratory Dewar Flask, 4.5 Liter Capacity, Stainless Steel
SDP
Catalog No. 501977013
Encompass
Dewar Flasks are designed for laboratory applications involving storage and transportation of liquid nitrogen and dry ice. Flasks feature double walled borosilicate glass, vacuum pumped and annealed interior for extra strength. Insulated, vented lid allows safe expelling of expelling gas. Lids clamp securely in place for convenient and safe transportation.
Catalog No. 50-197-7013 Supplier Research Products International Corp Supplier No. 280003
Dewar Flasks are designed for laboratory applications involving storage and transportation of liquid nitrogen and dry ice. Flasks feature double walled borosilicate glass, vacuum pumped and annealed interior for extra strength. Insulated, vented lid allows safe expelling of expelling gas. Lids clamp securely in place for convenient and safe transportation.