Learn More
Electron Microscopy Sciences Lab Drawer Organizer Lab Drawer Organizer, General
SDP
Catalog No. 5026832
Encompass_Preferred
We offer a variety of trays for your drawers or to fill in 6-Drawer Cart (64405-series) and 7-Shelf Cart (64406-series) above. These trays can store and sort supplies of all sizes and shapes. Trays are made of ABS, color white, and chemical resistant.
Catalog No. 50-268-32 Supplier Electron Microscopy Sciences Supplier No. 6437030
We offer a variety of trays for your drawers or to fill in 6-Drawer Cart (64405-series) and 7-Shelf Cart (64406-series) above. These trays can store and sort supplies of all sizes and shapes. Trays are made of ABS, color white, and chemical resistant.