Learn More
Electron Microscopy Sciences Lab Drawer Organizer Lab Drawer Organizer, Tray
SDP
Catalog No. 5026835
Encompass_Preferred
We offer a variety of trays for your drawers or to fill in 6-Drawer Cart (64405-series) and 7-Shelf Cart (64406-series) above. These trays can store and sort supplies of all sizes and shapes. Trays are made of ABS, color white, and chemical resistant. Divided into 16 equal sized compartments.
Catalog No. 50-268-35 Supplier Electron Microscopy Sciences Supplier No. 6437010
We offer a variety of trays for your drawers or to fill in 6-Drawer Cart (64405-series) and 7-Shelf Cart (64406-series) above. These trays can store and sort supplies of all sizes and shapes. Trays are made of ABS, color white, and chemical resistant. Divided into 16 equal sized compartments.