Learn More
Electron Microscopy Sciences Lab Drawer Organizer Lab Drawer Organizer, Utility
SDP
Catalog No. 5026834
Encompass_Preferred
We offer a variety of trays for your drawers or to fill in 6-Drawer Cart (64405-series) and 7-Shelf Cart (64406-series) above. These trays can store and sort supplies of all sizes and shapes. Trays are made of ABS, color white, and chemical resistant. For large items.
Catalog No. 50-268-34 Supplier Electron Microscopy Sciences Supplier No. 6437040
We offer a variety of trays for your drawers or to fill in 6-Drawer Cart (64405-series) and 7-Shelf Cart (64406-series) above. These trays can store and sort supplies of all sizes and shapes. Trays are made of ABS, color white, and chemical resistant. For large items.