Learn More
Accuflow Systems Saint-Gobain Chemfluor PTFE Film, 0.005 in, 6 in, 50 ft
Catalog No. NC2327706
Encompass_Preferred
Saint-Gobain Chemfluor PTFE fluoropolymer film is a premium-grade skived PTFE roll designed for laboratory and industrial lining, covering, and sealing applications. The film offers high chemical resistance and a non-stick surface suitable for scientific research and general-purpose protective uses.
  • Premium skived PTFE film made from virgin material.
  • 0.005 in thickness, 6 in width, 50 ft length.
  • High chemical resistance and non-stick surface.
  • Suitable for lining, covering, and sealing in lab and industrial environments.
  • Supplied in roll format for easy dispensing and cutting to size.
Catalog No. NC2327706 Supplier Accuflow Systems Supplier No. D1069278
Saint-Gobain Chemfluor PTFE fluoropolymer film is a premium-grade skived PTFE roll designed for laboratory and industrial lining, covering, and sealing applications. The film offers high chemical resistance and a non-stick surface suitable for scientific research and general-purpose protective uses.
  • Premium skived PTFE film made from virgin material.
  • 0.005 in thickness, 6 in width, 50 ft length.
  • High chemical resistance and non-stick surface.
  • Suitable for lining, covering, and sealing in lab and industrial environments.
  • Supplied in roll format for easy dispensing and cutting to size.