Learn More
Chemglass Life Sciences Lab Frame Foot
SDP
Catalog No. 50871987
Encompass_Preferred
Lab Frame Foot. Permanent lattice mounting to bench tops, floors or walls. Accommodates 1/2 (13mm) diameter rod. Supplied with three screws.
Catalog No. 50-871-987 Supplier Chemglass Life Sciences Supplier No. CG927401
Lab Frame Foot. Permanent lattice mounting to bench tops, floors or walls. Accommodates 1/2 (13mm) diameter rod. Supplied with three screws.