Learn More
Chemglass Life Sciences Lab Funnel, Disposable, Paper
SDP
Catalog No. 501311299
Encompass_Preferred
Lab Funnel, Disposable, Paper. • Made of clay coated, heavy paper stock• Use with virtually any liquid or powder• Helps reduce cross contamination• Good alternative to disposable plastic funnels• Customizable spout – trim to desired opening• Environmentally friendly – naturally biodegradableThe innovative “backrest” of these sturdy, paper funnels allows hands-free use when pouring viscous liquids. With a 1/4” opening, they fit a variety of vessels but are easily cut to any desired opening size or shape. Funnels stay in position with the application of a minimum amount of pressure. Use with virtually any liquid or powder but not recommended for hot liquids. Funnels are naturally biodegradable. Supplied flat to maintain cleanliness and facilitate storage.
Catalog No. 50-131-1299 Supplier Chemglass Life Sciences Supplier No. CLS2080100
Add to Cart
Add to Cart
Lab Funnel, Disposable, Paper. • Made of clay coated, heavy paper stock• Use with virtually any liquid or powder• Helps reduce cross contamination• Good alternative to disposable plastic funnels• Customizable spout – trim to desired opening• Environmentally friendly – naturally biodegradableThe innovative “backrest” of these sturdy, paper funnels allows hands-free use when pouring viscous liquids. With a 1/4” opening, they fit a variety of vessels but are easily cut to any desired opening size or shape. Funnels stay in position with the application of a minimum amount of pressure. Use with virtually any liquid or powder but not recommended for hot liquids. Funnels are naturally biodegradable. Supplied flat to maintain cleanliness and facilitate storage.