Learn More
Bellco Glass, Inc. Lab Grab MultiUse
SDP
Catalog No. 502330138
Encompass_Preferred
CELLTREAT Lab Grab allows handsfree and easy grasping of small items as well as hot or cold samples. Large extension radius to pick up small to midsized objects including cryogenic vials screw top micro tubes micro centrifuge tubes and cell strainers.Specifications Retraction feature aids in stability and strength of grip Textured handle prevents slipping when wearing gloves Allmetal construction allows autoclaving Case Quantity 2
Catalog No. 50-233-0138 Supplier Bellco Glass, Inc. Supplier No. 230110000
CELLTREAT Lab Grab allows handsfree and easy grasping of small items as well as hot or cold samples. Large extension radius to pick up small to midsized objects including cryogenic vials screw top micro tubes micro centrifuge tubes and cell strainers.Specifications Retraction feature aids in stability and strength of grip Textured handle prevents slipping when wearing gloves Allmetal construction allows autoclaving Case Quantity 2