Learn More
PHCbi 22.0 cu.ft. -30°C Upright Laboratory Grade Freezer
Catalog No. 502720058
Encompass_Preferred
PHCbi High Performance Biomedical Freezers are designed for short- or intermediate-term storage at temperatures as low as -30°C. Constructed with high performance laboratory and clinical grade refrigeration systems, these freezers are used in medical, biotechnology and industrial labs for storage of blood components, enzymes, culture media, reagents, specimens and vaccines.The MDF-U731-PA -30°C Upright, Medical-Grade Freezer was designed with the features to give superior uniformity, microprocessor control and large storage capacity. Two access ports, casters, remote alarm contacts and security features are included as standard features.Effective capacity: 22.0 cu.ft. (623 L)Temperature range: -15℃ to -30℃
Catalog No. 50-272-0058 Supplier PHCbi Supplier No. MDFU731PA
Add to Cart
Add to Cart
Request a Quote
PHCbi High Performance Biomedical Freezers are designed for short- or intermediate-term storage at temperatures as low as -30°C. Constructed with high performance laboratory and clinical grade refrigeration systems, these freezers are used in medical, biotechnology and industrial labs for storage of blood components, enzymes, culture media, reagents, specimens and vaccines.The MDF-U731-PA -30°C Upright, Medical-Grade Freezer was designed with the features to give superior uniformity, microprocessor control and large storage capacity. Two access ports, casters, remote alarm contacts and security features are included as standard features.Effective capacity: 22.0 cu.ft. (623 L)Temperature range: -15℃ to -30℃