Learn More
Cole-Parmer Lab-Grade Glass Dewar Flask, vented polyethylene stopper, 350 mL
Catalog No. NC1532803 Shop All Cole-Parmer Products
Encompass
Highly efficient laboratory-grade Dewars feature low heat conductivity and high vacuum. Annealed borosilicate glass reduces risk of flask implosion. Weighted base reduces spillage. Resilient plastic mesh covering cushions the flask and ensures a firm grip. Vented polyethylene stopper (included) minimizes evaporation.
Catalog No. NC1532803 Supplier Cole-Parmer Supplier No. 0376610
Highly efficient laboratory-grade Dewars feature low heat conductivity and high vacuum. Annealed borosilicate glass reduces risk of flask implosion. Weighted base reduces spillage. Resilient plastic mesh covering cushions the flask and ensures a firm grip. Vented polyethylene stopper (included) minimizes evaporation.