Learn More
Electron Microscopy Sciences Lab Island™ Storage Cart Model B
SDP
Catalog No. 501901897
Encompass_Preferred
This mobility storage cart is the perfect unit for your lab. Made of HDPE, it comes with the roll top door offering additional safety over traditional doors. Features one large pull-out shelf with 1.5" safety lip made entirely from HDPE. Secure lock door with two keys and lockable caster wheels. Translucense of HDPE allows for easy viewing of contents inside.
Catalog No. 50-190-1897 Supplier Electron Microscopy Sciences Supplier No. 6440020
This mobility storage cart is the perfect unit for your lab. Made of HDPE, it comes with the roll top door offering additional safety over traditional doors. Features one large pull-out shelf with 1.5" safety lip made entirely from HDPE. Secure lock door with two keys and lockable caster wheels. Translucense of HDPE allows for easy viewing of contents inside.