Learn More
Med Vet International Premiere Lab Jack
Catalog No. 503046922
Encompass_Preferred
This Premiere brand lab jack is designed to support and elevate beakers, flasks, and other lab equipment that may be too hot or heavy to lift. 5 1/2? x 5 3/4? surface platform elevates from 3? to 10 1/2? by simple cranking of easy grip knob. Sturdy nickel plated steel can support up to 100 lbs.
Catalog No. 50-304-6922 Supplier Med Vet International Supplier No. 975701
This Premiere brand lab jack is designed to support and elevate beakers, flasks, and other lab equipment that may be too hot or heavy to lift. 5 1/2? x 5 3/4? surface platform elevates from 3? to 10 1/2? by simple cranking of easy grip knob. Sturdy nickel plated steel can support up to 100 lbs.