Learn More
Electron Microscopy Sciences Lab Jacks
SDP
Catalog No. 5031302
Encompass_Preferred
This LabJack provides stable height adjustment and support for items in the lab such as flasks, baths or hot plate stirrers. Top and bottom plates are constructed of anodized aluminum while the support mechanism is made of chrome plated steel.
Catalog No. 50-313-02 Supplier Electron Microscopy Sciences Supplier No. 6322524
This LabJack provides stable height adjustment and support for items in the lab such as flasks, baths or hot plate stirrers. Top and bottom plates are constructed of anodized aluminum while the support mechanism is made of chrome plated steel.