Learn More
Braintree Scientific Lab Light
SDP
Catalog No. 501955007
Encompass
Lab Light - Unique magnetic base/puck system holds the lab light firmly on the base in any location. Magnetic base/puck system allows the lab light to attach to non-metallic surfaces. Plug the lab light into the On/Off Plug Switch for easy on/off operation. LED on 10 Flex Arm. Ultra Strong Magnetic Base/Puck System to Anchor Light on Non-Metallic Surfaces. Attaches Directly to Metallic Surfaces. Universal Power Supply (110-220 V). Small Base (1 dia.) **Sold for research purposes only and not for human use**
Catalog No. 50-195-5007 Supplier Braintree Scientific Supplier No. LL13
Lab Light - Unique magnetic base/puck system holds the lab light firmly on the base in any location. Magnetic base/puck system allows the lab light to attach to non-metallic surfaces. Plug the lab light into the On/Off Plug Switch for easy on/off operation. LED on 10 Flex Arm. Ultra Strong Magnetic Base/Puck System to Anchor Light on Non-Metallic Surfaces. Attaches Directly to Metallic Surfaces. Universal Power Supply (110-220 V). Small Base (1 dia.) **Sold for research purposes only and not for human use**