Learn More
Grainger Laboratory Liner Wh Pp 24X24 Pk10
Catalog No. NC9978559
Encompass_Preferred
Liner, Bench; Lab Safety Supply; Polypropylene made; White, Protect benchtops, Shelves and drawers from hot/cold spills, Polypropylene resists Temperatures from 29 to 230 deg.; Superior chemical resistance, Autoclavable; Size: 24 x 24 in.
Catalog No. NC9978559 Supplier Grainger Supplier No. 9EKP5
Liner, Bench; Lab Safety Supply; Polypropylene made; White, Protect benchtops, Shelves and drawers from hot/cold spills, Polypropylene resists Temperatures from 29 to 230 deg.; Superior chemical resistance, Autoclavable; Size: 24 x 24 in.