Learn More
Adam Equipment LAB 254i Luna Analytical Balance 250g capacity x 0.0001g readability
Supplier:  Adam Equipment LAB 254I
Encompass_Preferred
Adam Equipment BCT Label Printing Scales feature a removable, easy-to-clean stainless steel platform, BCT advanced label printing scales provide excellent value for industrial counting and weighing tasks. The BCT features a printer that can print weight, QR codes, bar codes and labels, which can be customized using the included software. Store and recall up to 1006 PLUs or piece weights to improve efficiency in production or warehouse applications. RS-232 interface provide speedy connection to computers and printers. BCT offers a choice of two platform sizes, making it ideal for counting inventory, shipping and receiving, and batching items for processing. With an easy-to-clean stainless steel platform, BCT scales offer excellent value for industrial counting tasks. BCT contains a printer that prints QR codes, bar codes and labels. Store up to 1006 PLUs or piece weights to expedite inventory and batching jobs. Rechargeable battery included for operation.
Catalog No. NC2611810