Learn More
Daigger & Company Black Lab Marker, 10/pack
SDP
Catalog No. NC0623278
Encompass
Lab Markers contain permanent, quick-drying ink that writes on cold or wet surfaces, plastic bags, or disposable labware. Securline Lab Markers will not smear on reusable glass, metal or porcelain labware, but can be scrubbed off. Fine tip. Package of 10.
Catalog No. NC0623278 Supplier Daigger & Company Supplier No. PDY140020PDC
Lab Markers contain permanent, quick-drying ink that writes on cold or wet surfaces, plastic bags, or disposable labware. Securline Lab Markers will not smear on reusable glass, metal or porcelain labware, but can be scrubbed off. Fine tip. Package of 10.