Learn More
Electron Microscopy Sciences Laboratory Notebook Lined 200 pages, Red
SDP
Catalog No. 501929476
Encompass_Preferred
Keep your information safe and well organized with these notebooks. Use for recording research data, experimental procedures, and design ideas.
Hardbound book with waterproof and chemical-resistant imitation leather cover.
Features a foil stamped title: LABORATORY NOTEBOOK.
Numbered, acid-free, pages include spaces for title, project number, book number, date, and witness signature.
Red ribbon marker for quick and easy access to your notes.
All notebooks contain periodic and conversion tables in the back.
Measures: 8.58" x 11.06" (218 x 281 mm). College-ruled pages for easy report writing.
Color: Red
Catalog No. 50-192-9476 Supplier Electron Microscopy Sciences Supplier No. 3802540
Keep your information safe and well organized with these notebooks. Use for recording research data, experimental procedures, and design ideas.
Hardbound book with waterproof and chemical-resistant imitation leather cover.
Features a foil stamped title: LABORATORY NOTEBOOK.
Numbered, acid-free, pages include spaces for title, project number, book number, date, and witness signature.
Red ribbon marker for quick and easy access to your notes.
All notebooks contain periodic and conversion tables in the back.
Measures: 8.58" x 11.06" (218 x 281 mm). College-ruled pages for easy report writing.
Color: Red