Learn More
Description
- Drum is constructed of durable HDPE
- Nestable storage to optimize shipping and storage space
- Ideal for use with smaller non-damaged bottles, cans, carboys and non-leaking hazardous materials
- Available with various closure types including plastic or metal lever locking bands, or bolt rings
- Food grade quality, FDA 21 CFR 177.1520
- Operating Temp range (-30°F to 130°F)
- Drop tested-max weight @ 0°F (48 hr.)
- Flammability Rating: UL 94HB
- If used with leaking containers, must pack with sufficient sorbents
- No free liquids
Specifications
Specifications
| Product Type | Drum |
| Closure Included | Yes |
| Closure Type | Plastic Lever-lock |
| Capacity (English) | 10 gal. |
| Capacity (Metric) | 37.9 L |
| Color | Yellow |
| For Use With (Application) | Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration |
| Material | HDPE |
| Head Style | Open |
| Max. Temperature (Metric) | 54.4°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
