Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lab Performance Check, Method 200.7, Ricca Chemical
SDP

Catalog No. RLPC1500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RLPC1-500 500 mL
RLPC1-100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RLPC1-500 Supplier Ricca Chemical Company Supplier No. RLPC1500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Amber Poly Bottle
CAS Min % 91.0, 6.5, 0.05, 0.03, 0.02, 0.01
CAS Max % 94.0, 6.76, 0.05, 0.03, 0.02, 0.02
CAS 7697-37-2, 16919-19-0, 7722-76-1, 7784-27-2, 584-08-7
Methods Method 200.7
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.