Learn More
Cole-Parmer Oakton General-Purpose pH Probe, Refillable, DJ, Glass, 3 ft Cable, BNC
Catalog No. NC9051843 Shop All Cole-Parmer Products
Encompass_Preferred
Compatible with all Oakton brand pH meters. Ideal for general purpose measurements. Withstands aggressive chemicals. Double junction type. Oakton combination electrodes are available in a variety of styles to meet the needs of your application. Epoxy electrodes are impact resistant and provide excellent durability for field applications. Glass electrodes withstand aggressive chemicals. Choose junction and tip style based on the needs of your application. All electrodes feature Ag/AgCl reference cells. Includes: Direct-connect models include a BNC connector.
Catalog No. NC9051843 Supplier Cole-Parmer Supplier No. 3580504
Compatible with all Oakton brand pH meters. Ideal for general purpose measurements. Withstands aggressive chemicals. Double junction type. Oakton combination electrodes are available in a variety of styles to meet the needs of your application. Epoxy electrodes are impact resistant and provide excellent durability for field applications. Glass electrodes withstand aggressive chemicals. Choose junction and tip style based on the needs of your application. All electrodes feature Ag/AgCl reference cells. Includes: Direct-connect models include a BNC connector.