Learn More
Research Products International Corp Lab Roller Rack for 10x15ml Tubes
SDP
Catalog No. 501978212
Encompass
Lab Roller Rack for use with Lab Roller Tube Rotator. Rack holds 10 x 15 ml (16 - 17 mm) and 12 x 5 ml (12 - 13 mm) tubes or two microplates.
Catalog No. 50-197-8212 Supplier Research Products International Corp Supplier No. 248141
Add to Cart
Add to Cart
Lab Roller Rack for use with Lab Roller Tube Rotator. Rack holds 10 x 15 ml (16 - 17 mm) and 12 x 5 ml (12 - 13 mm) tubes or two microplates.