Learn More
Research Products International Corp Laboratory Grade Rubber Stoppers w/Hole, BLK SBR Rubber, Size #8 1/2, 43 x 36 x 25mm, 10 per Package
SDP
Catalog No. 502229720
Encompass
Laboratory Grade Rubber Stoppers with hole are offered in a wide range of common sizes for most laboratory applications. Available standard black SBR rubber. Material can typically withstand temperatures up to about 100°C (212°F). Rubber stoppers with hole are molded in the USA according to standard industry size specification. 
Packaged in approximately one pound packages.
Catalog No. 50-222-9720 Supplier Research Products International Corp Supplier No. 259150H85
Laboratory Grade Rubber Stoppers with hole are offered in a wide range of common sizes for most laboratory applications. Available standard black SBR rubber. Material can typically withstand temperatures up to about 100°C (212°F). Rubber stoppers with hole are molded in the USA according to standard industry size specification. 
Packaged in approximately one pound packages.