Learn More
Electron Microscopy Sciences AMT Series Laboratory Side Work Table, 120 x 75 cm, Stainless Steel Surface
SDP
Catalog No. 501930088
Encompass_Preferred
These laboratory tables feature a rigid frame with levelling feet which ensure the table is in excellent contact with the floor. Available with White Epoxy, Stainless Steel or Trespa top. Options include : monitor arms and castor feet.
Size: 120 x 75 cm
Surface: Stainless Steel
Catalog No. 50-193-0088 Supplier Electron Microscopy Sciences Supplier No. 8920
These laboratory tables feature a rigid frame with levelling feet which ensure the table is in excellent contact with the floor. Available with White Epoxy, Stainless Steel or Trespa top. Options include : monitor arms and castor feet.
Size: 120 x 75 cm
Surface: Stainless Steel