Learn More
Andwin Scientific LAB SOAKERS 20X150 ROLLS CS/2
SDP
Catalog No. NC9773564
Encompass_Preferred
The Nalgene Standard Versi-Dry Paper Lab Soaker Roll is able to absorb 750 mL per square meter. This product features a skid-resistant waterproof chemical-resistant polyethylene backing that allows easier spill containment. The soaker can also be utilized as a tray drawer or shelf liner or to wipe up spills. This measures 20 inch wide x 150 foot long. It is sold in a case of 2.
Catalog No. NC9773564 Supplier Andwin Scientific Supplier No. 62065
The Nalgene Standard Versi-Dry Paper Lab Soaker Roll is able to absorb 750 mL per square meter. This product features a skid-resistant waterproof chemical-resistant polyethylene backing that allows easier spill containment. The soaker can also be utilized as a tray drawer or shelf liner or to wipe up spills. This measures 20 inch wide x 150 foot long. It is sold in a case of 2.