Learn More
Polysciences, Inc. L.A.B. Solution (Liberate Antibody Binding Solution), 500 mL
SDP
Catalog No. NC9109007
Encompass_Preferred
L.A.B. Solution liberates your antibody binding sites for improved immunohistochemical (IHC) staining.
Catalog No. NC9109007 Supplier Polysciences, Inc. Supplier No. 24310500
L.A.B. Solution liberates your antibody binding sites for improved immunohistochemical (IHC) staining.