Learn More
Electron Microscopy Sciences Lab-Sorb 16 oz jar 16 OZ
SDP
Catalog No. 5034509
Encompass_Preferred
Lab-Sorb converts liquid waste to solid waste for easy disposal. LabSorb liquids to solids in seconds and the granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste. It reduces the possibility of accidentally spilling liquid waste in the lab and if spills do occur, use LabSorb for safe liquid containment during cleanup. Apply to any volume of liquid waste including 500 ml, 600 ml, 1L and 2L vessels. One jar of LabSorb can absorb up to 50 litres of liquid.
Catalog No. 50-345-09 Supplier Electron Microscopy Sciences Supplier No. 6855206
Lab-Sorb converts liquid waste to solid waste for easy disposal. LabSorb liquids to solids in seconds and the granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste. It reduces the possibility of accidentally spilling liquid waste in the lab and if spills do occur, use LabSorb for safe liquid containment during cleanup. Apply to any volume of liquid waste including 500 ml, 600 ml, 1L and 2L vessels. One jar of LabSorb can absorb up to 50 litres of liquid.